missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyne, 98%

Catalog No. AC329905000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329905000 500 mL Glass bottle
AC329900250 25 mL Glass bottle
AC329901000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC329905000 Supplier Thermo Scientific Chemicals Supplier No. 329905000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 693-02-7 | C6H10 | 82.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 693-02-7
Molecular Formula C6H10
Molecular Weight (g/mol) 82.13
InChI Key CGHIBGNXEGJPQZ-UHFFFAOYSA-N
Synonym 1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
PubChem CID 12732
IUPAC Name hex-1-yne
SMILES CCCCC#C

Specifications

Melting Point -132.0°C
Color Colorless
Density 0.7100g/mL
Boiling Point 71.0°C to 72.0°C
Flash Point −021°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3974
Linear Formula CH3(CH2)3C≡CH
Quantity 500 mL
Beilstein 01,253
Specific Gravity 0.71
Formula Weight 82.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Hexyne
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 211-736-9

RUO – Research Use Only