missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexyl)triphenylphosphonium bromide, 98%

Catalog No. AAA1382614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1382614 25 g
AAA1382622 100 g
AAA1382636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1382614 Supplier Thermo Scientific Chemicals Supplier No. A1382614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4762-26-9 | C24H28BrP | 427.37 g/mol

(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store under inert gas. Protect from light. Store away from oxidizing materials.

Chemical Identifiers

CAS 4762-26-9
Molecular Formula C24H28BrP
Molecular Weight (g/mol) 427.37
MDL Number MFCD00011860
InChI Key PWDFZWZPWFYFTC-UHFFFAOYSA-M
Synonym hexyltriphenylphosphonium bromide, n-hexyl-triphenylphosphonium bromide, phosphonium, hexyltriphenyl-, bromide, hexyltriphenylphosphoniumbromide, hexyltriphenylphosphanium bromide, n-hexyl triphenylphosphonium bromide, c24h28p.br, acmc-209k9x, n-hexyltriphenylphosphonium bromide, hexyl triphenyl phosphanium bromide
PubChem CID 2724569
SMILES [Br-].CCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 199°C to 202°C
Quantity 25 g
Beilstein 3580731
Sensitivity Hygroscopic
Solubility Information Soluble in water and methanol.
IUPAC Name hexyltriphenylphosphanium bromide
Formula Weight 427.37
Percent Purity 98%
Chemical Name or Material (1-Hexyl)triphenylphosphonium bromide
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-301-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only