missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl-3-methylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. H11005G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H11005G Supplier TCI America Supplier No. H11005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 460345-16-8
Molecular Formula C11H19F3N2O3S
Molecular Weight (g/mol) 316.339
MDL Number MFCD03427621
InChI Key RABFGPMWVQNDHI-UHFFFAOYSA-M
Synonym 1-hexyl-3-methylimidazolium trifluoromethanesulfonate, 1-hexyl-3-methylimidazolium trifluoromethansulfonate, 1-hexyl-3-methylimidazolium triflate, hmim otf, dsstox_cid_27847, dsstox_rid_82600, dsstox_gsid_47871, ksc491c7h, 1-hexyl-3-methylimidazoliumtrifluoromethansulfonate, 1-hexyl-3-methyl-imidazol-3-ium; trifluoromethanesulfonate
PubChem CID 2734247
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 316.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Hexyl-3-methylimidazolium Trifluoromethanesulfonate