missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl-3-methylimidazolium Chloride 98.0+%, TCI America™

$80.16 - $231.14

Chemical Identifiers

CAS 171058-17-6
Molecular Formula C10H19N2
Molecular Weight (g/mol) 167.28
MDL Number MFCD03093289
InChI Key RVEJOWGVUQQIIZ-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f
PubChem CID 2734163
ChEBI CHEBI:61340
IUPAC Name 3-hexyl-1-methyl-1H-imidazol-3-ium
SMILES CCCCCC[N+]1=CN(C)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
H10975G
View Documents
TCI America
H10975G
5 g
Each for $80.16
Only null left
 
H109725G
View Documents
TCI America
H109725G
25 g
Each for $231.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

171058-17-6
167.28
RVEJOWGVUQQIIZ-UHFFFAOYSA-N
2734163
3-hexyl-1-methyl-1H-imidazol-3-ium
C10H19N2
MFCD03093289
1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f
CHEBI:61340
CCCCCC[N+]1=CN(C)C=C1

Specifications

171058-17-6
C10H19N2
5 g
RVEJOWGVUQQIIZ-UHFFFAOYSA-N
3-hexyl-1-methyl-1H-imidazol-3-ium
2734163
202.73
Liquid
Yellow
MFCD03093289
1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f
CCCCCC[N+]1=CN(C)C=C1
167.28
CHEBI:61340
≥98.0% (T)
1-Hexyl-3-methylimidazolium Chloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents