accessibility menu, dialog, popup

UserName

H3CNNCH3

CAS RN 171058-17-6

IUPAC Name: 3-hexyl-1-methyl-1H-imidazol-3-ium
Synonyms: 1-hexyl-3-methylimidazolium
Molecular Weight (g/mol): 167.28
Molecular Formula: C10H19N2
InChi Key: RVEJOWGVUQQIIZ-UHFFFAOYSA-N
SMILES: CCCCCC[N+]1=CN(C)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)