missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
$165.90 - $486.52
Chemical Identifiers
| CAS | 382150-50-7 |
|---|---|
| Molecular Formula | C12H19F6N3O4S2 |
| Molecular Weight (g/mol) | 447.411 |
| MDL Number | MFCD08458931 |
| InChI Key | RCNFOZUBFOFJKZ-UHFFFAOYSA-N |
| PubChem CID | 10411334 |
| IUPAC Name | bis(trifluoromethylsulfonyl)azanide;1-hexyl-3-methylimidazol-3-ium |
| SMILES | CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Chemical Identifiers
| 382150-50-7 | |
| 447.411 | |
| RCNFOZUBFOFJKZ-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-hexyl-3-methylimidazol-3-ium |
| C12H19F6N3O4S2 | |
| MFCD08458931 | |
| 10411334 | |
| CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Specifications
| 382150-50-7 | |
| Yellow | |
| MFCD08458931 | |
| RCNFOZUBFOFJKZ-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-hexyl-3-methylimidazol-3-ium | |
| 10411334 | |
| ≥98.0% (HPLC,T) | |
| 1-Hexyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide |
| -9°C | |
| C12H19F6N3O4S2 | |
| 5 g | |
| CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 447.411 | |
| 447.41 | |
| Liquid |
Safety and Handling
TSCA : No