CAS RN 382150-50-7
IUPAC Name:
1-hexyl-3-methyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms:
1-hexyl-3-methylimidazolium; bistriflylimide anion
Molecular Weight (g/mol):
447.41
Molecular Formula:
C12H19F6N3O4S2
InChi Key:
RCNFOZUBFOFJKZ-UHFFFAOYSA-N
SMILES:
CCCCCCN1C=C[N+](C)=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
1-Hexyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
CAS: 382150-50-7 Molecular Formula: C12H19F6N3O4S2 Molecular Weight (g/mol): 447.411 MDL Number: MFCD08458931 InChI Key: RCNFOZUBFOFJKZ-UHFFFAOYSA-N PubChem CID: 10411334 IUPAC Name: bis(trifluoromethylsulfonyl)azanide;1-hexyl-3-methylimidazol-3-ium SMILES: CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F