missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. H14235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14235G 5 g
H142325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14235G Supplier TCI America Supplier No. H14235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 382150-50-7
Molecular Formula C12H19F6N3O4S2
Molecular Weight (g/mol) 447.411
MDL Number MFCD08458931
InChI Key RCNFOZUBFOFJKZ-UHFFFAOYSA-N
PubChem CID 10411334
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-hexyl-3-methylimidazol-3-ium
SMILES CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point -9°C
Color Yellow
Quantity 5 g
Formula Weight 447.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Hexyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide