missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanol, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020401
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020401 1 mL
111018781 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020401 Supplier Merck Emd Millipore Supplier No. 731171MLF
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 559°F
CAS 111-27-3
Vapor Pressure 1 mmHg (25.6°C)
Vapor Density 4.5 (vs. air)
Density 0.814 g/mL (at 25°C)
Flash Point 60°C
Physical Form Neat
Linear Formula CH3(CH2)5 OH
MDL Number MFCD00002982
Melting Point -52°C
Boiling Point 156°C to 157°C
Synonym Hexyl alcohol
Type Analytical Standard
Quantity 1 mL
Formula Weight 102.17
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less