Learn More
1-Hexanethiol, 96%
CAS: 111-31-9 | C6H14S | 118.24 g/mol
$74.32 - $180.05
Chemical Identifiers
| CAS | 111-31-9 |
|---|---|
| Molecular Formula | C6H14S |
| Molecular Weight (g/mol) | 118.24 |
| MDL Number | MFCD00004909 |
| InChI Key | PMBXCGGQNSVESQ-UHFFFAOYSA-N |
| Synonym | 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan |
| PubChem CID | 8106 |
| IUPAC Name | hexane-1-thiol |
| SMILES | CCCCCCS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL09659AC
|
Thermo Scientific Chemicals
L09659AC |
25 mL |
N/A
|
|
|||||
|
AAL09659AE
|
Thermo Scientific Chemicals
L09659AE |
100 mL |
N/A
|
|
|||||
Description
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1Â Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2Â to provide a clean Cu surface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1Â Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2Â to provide a clean Cu surface.
Solubility
Difficult to mix in water.
Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 111-31-9 | |
| 118.24 | |
| PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
| 8106 | |
| CCCCCCS |
| C6H14S | |
| MFCD00004909 | |
| 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
| hexane-1-thiol |
Specifications
| 111-31-9 | |
| 0.838 | |
| 30°C (86°F) | |
| C6H14S | |
| CH3(CH2)5SH | |
| 25 mL | |
| 1731295 | |
| 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
| PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
| hexane-1-thiol | |
| 8106 | |
| 97% |
| -81°C | |
| 150°C to 154°C | |
| Strong | |
| 1.448 | |
| MFCD00004909 | |
| UN3071 | |
| Air Sensitive | |
| Difficult to mix in water. | |
| CCCCCCS | |
| 118.24 | |
| 118.24 | |
| 1-Hexanethiol |
Safety and Handling
GHS H Statement
H301-H331-H226
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P330-P370+P378q-P501c
H226-H302-H331
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 203-857-0
RTECSNumber : MO4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only