missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanethiol, 96%

Catalog No. AAL09659AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL09659AC 25 mL
AAL09659AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL09659AC Supplier Thermo Scientific Chemicals Supplier No. L09659AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-31-9 | C6H14S | 118.24 g/mol

It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

Solubility
Difficult to mix in water.

Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 111-31-9
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00004909
InChI Key PMBXCGGQNSVESQ-UHFFFAOYSA-N
Synonym 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
PubChem CID 8106
IUPAC Name hexane-1-thiol
SMILES CCCCCCS

Specifications

Melting Point -81°C
Density 0.838
Boiling Point 150°C to 154°C
Flash Point 30°C (86°F)
Odor Strong
Refractive Index 1.448
Linear Formula CH3(CH2)5SH
Quantity 25 mL
UN Number UN3071
Beilstein 1731295
Sensitivity Air Sensitive
Solubility Information Difficult to mix in water.
Formula Weight 118.24
Percent Purity 97%
Chemical Name or Material 1-Hexanethiol
Show More Show Less
Hazard Category H226-H302-H331
Hazard Statement GHS H Statement
H301-H331-H226
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P330-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-857-0
RTECSNumber MO4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only