missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic Acid Sodium Salt, Monohydrate, 98%, Spectrum™ Chemical

Catalog No. 18608113
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608113 5 g Amber Glass Bottle
18608114 25 g Amber Glass Bottle
18608115 100 g Amber Glass Bottle
18608116 1 kg Amber Glass Bottle
18608117 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608113 Supplier Spectrum Chemical Mfg Cor Supplier No. H10325GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 207300-91-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.