Learn More
1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade
CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol
Supplier: Thermo Scientific Chemicals 411601000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatographySpecifications
| 1-Hexanesulfonic acid, sodium salt hydrate | |
| (0.25M aq. soln.) (1cm cell vs HPLC grade water), 0.2 max. at 210nm, 0.1 max. at 220nm, 0.075 max. at 230nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.04 max. at 254nm, 0.04 max. at 280nm, (0.25 M aq. soln.) (1 cm cell vs HPLC grade water), (0.25 M aq. soln.) (1 cm cell vs HPLC grade water) at 0nm | |
| Negligible | |
| 98% | |
| C6H13NaO3S | |
| MFCD00007542 | |
| 04, IV, 54 | |
| Solubility in water: Soluble | |
| [Na+].CCCCCCS([O-])(=O)=O | |
| 188.22 | |
| 188.22 | |
| ≥97.5% (on dry substance) |
| 2832-45-3 | |
| Colorless to Yellow | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)5SO3Na·xH2O | |
| 100 g | |
| sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
| QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
| sodium;hexane-1-sulfonate | |
| 23677630 | |
| Negligible | |
| Liquid |
Chemical Identifiers
| 2832-45-3 | |
| 188.22 | |
| QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
| 23677630 | |
| [Na+].CCCCCCS([O-])(=O)=O |
| C6H13NaO3S | |
| MFCD00007542 | |
| sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
| sodium;hexane-1-sulfonate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only