missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade

CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol

Supplier:  Thermo Scientific Chemicals 411600250

 View more versions of this product

Catalog No. AC411600250

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
Specifications

Specifications

1-Hexanesulfonic acid, sodium salt hydrate
(0.25M aq. soln.) (1cm cell vs HPLC grade water), 0.2 max. at 210nm, 0.1 max. at 220nm, 0.075 max. at 230nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.04 max. at 254nm, 0.04 max. at 280nm, (0.25 M aq. soln.) (1 cm cell vs HPLC grade water), (0.25 M aq. soln.) (1 cm cell vs HPLC grade water) at 0nm
Colorless to Yellow
Authentic
Glass bottle
CH3(CH2)5SO3Na·xH2O
25 g
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium;hexane-1-sulfonate
23677630
Negligible
Liquid
2832-45-3
>300°C
Negligible
98%
C6H13NaO3S
MFCD00007542
04, IV, 54
(10% in water) Clear colorless
[Na+].CCCCCCS([O-])(=O)=O
188.22
188.22
98%

Chemical Identifiers

2832-45-3
188.22
QWSZRRAAFHGKCH-UHFFFAOYSA-M
23677630
[Na+].CCCCCCS([O-])(=O)=O
C6H13NaO3S
MFCD00007542
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
sodium;hexane-1-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only