missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic Acid Sodium Salt (HPLC), Fisher Chemical™

Catalog No. p-8820000
Change view
Click to view available options
Quantity:
100 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O307225 25 g
O3072100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O307225 Supplier Thermo Fisher Scientific Supplier No. O3072 25
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improve HPLC separation of basic analytes by ion pairing.

  • High purity
  • Application tested
  • Dissolves easily

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
IUPAC Name sodium hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

Alpha Vector Hexanesulfonic Acid Sodium Salt
Absorbance Pass Test
Melting Point 300°C
Color White
Assay Percent Range ≥98 %
Packaging Glass Bottle
Linear Formula CH3(CH2)5SO3Na
Quantity 25 g
Solubility Information Soluble in water
Formula Weight 188.22
Percent Purity ≥98%
Grade HPLC
Physical Form Powder or Solid
Chemical Name or Material 1-Hexanesulfonic Acid Sodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0