Learn More
1-Hexadecylamine, tech. 90%, remainder mainly 1-octadecylamine
CAS: 143-27-1 | C16H35N | 241.46 g/mol
$64.60 - $243.62
Chemical Identifiers
| CAS | 143-27-1 |
|---|---|
| Molecular Formula | C16H35N |
| Molecular Weight (g/mol) | 241.46 |
| MDL Number | MFCD00008158 |
| InChI Key | FJLUATLTXUNBOT-UHFFFAOYSA-N |
| Synonym | hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine |
| PubChem CID | 8926 |
| IUPAC Name | hexadecan-1-amine |
| SMILES | CCCCCCCCCCCCCCCCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2245914
|
Thermo Scientific Chemicals
B2245914 |
25 g |
Each for $64.60
|
|
|||||
|
AAB2245930
|
Thermo Scientific Chemicals
B2245930 |
250 g |
Each for $86.86
|
|
|||||
|
AAB224590B
|
Thermo Scientific Chemicals
B224590B |
1000 g |
Each for $243.62
|
|
|||||
Description
1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents, acids.
Chemical Identifiers
| 143-27-1 | |
| 241.46 | |
| FJLUATLTXUNBOT-UHFFFAOYSA-N | |
| 8926 | |
| CCCCCCCCCCCCCCCCN |
| C16H35N | |
| MFCD00008158 | |
| hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine | |
| hexadecan-1-amine |
Specifications
| 143-27-1 | |
| 0.813 | |
| 140°C (284°F) | |
| C16H35N | |
| MFCD00008158 | |
| UN3259 | |
| hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine | |
| FJLUATLTXUNBOT-UHFFFAOYSA-N | |
| hexadecan-1-amine | |
| 8926 | |
| 90% | |
| 1-Hexadecylamine |
| 42°C to 47°C | |
| 144°C (2 mmHg) | |
| 90% | |
| 1.4496 | |
| 25 g | |
| 1634065 | |
| Insoluble in water. | |
| CCCCCCCCCCCCCCCCN | |
| 241.46 | |
| 241.46 | |
| Technical | |
| remainder mainly 1-octadecylamine |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 205-596-8
RTECSNumber : ML9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only