missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecylamine, 90%

CAS: 143-27-1 | C16H35N | 241.46 g/mol

$51.28 - $114.06

Chemical Identifiers

CAS 143-27-1
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
MDL Number MFCD00008158
InChI Key FJLUATLTXUNBOT-UHFFFAOYSA-N
Synonym hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine
PubChem CID 8926
IUPAC Name hexadecan-1-amine
SMILES CCCCCCCCCCCCCCCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120511000
SDS View Documents
Thermo Scientific Chemicals
120511000
100 g Glass bottle
Each for $51.28
Only null left
 
AC120515000
SDS View Documents
Thermo Scientific Chemicals
120515000
500 g Glass bottle
Each for $114.06
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

143-27-1
241.46
FJLUATLTXUNBOT-UHFFFAOYSA-N
8926
CCCCCCCCCCCCCCCCN
C16H35N
MFCD00008158
hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine
hexadecan-1-amine

Specifications

143-27-1
White
140°C
88% min. (GC)
C16H35N
MFCD00008158
04,202
Solubility in water: insoluble
CCCCCCCCCCCCCCCCN
241.46
241.46
Low Melting Crystalline Mass or Chunks
38.0°C to 47.0°C
330.0°C
Authentic
Glass bottle
CH3(CH2)15NH2
100 g
hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine
FJLUATLTXUNBOT-UHFFFAOYSA-N
hexadecan-1-amine
8926
90%
1-Hexadecylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 205-596-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only