Learn More
(1-Hexadecyl)triphenylphosphonium bromide, 98+%
CAS: 14866-43-4 | C34H48BrP | 567.64 g/mol
$72.38 - $720.25
Chemical Identifiers
| CAS | 14866-43-4 |
|---|---|
| Molecular Formula | C34H48BrP |
| Molecular Weight (g/mol) | 567.64 |
| MDL Number | MFCD00051858 |
| InChI Key | UXMZNEHSMYESLH-UHFFFAOYSA-M |
| Synonym | hexadecyltriphenylphosphonium bromide, 1-hexadecyl triphenylphosphonium bromide, cetyltriphenylphosphonium bromide, phosphonium, hexadecyltriphenyl-, bromide, hexadecyl triphenyl phosphanium bromide, hexadecyltriphenylphosphanium bromide, acmc-1boiz, hexadecyl triphenyl phosphonium bromide, hexadecyl triphenyl phosphonium chloride, 1-hexadecyl triphenylphosphoniumbromide |
| PubChem CID | 3084210 |
| IUPAC Name | hexadecyl(triphenyl)phosphanium;bromide |
| SMILES | [Br-].CCCCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAA1518006
|
Thermo Scientific Chemicals
A1518006 |
5 g |
Each for $72.38
|
|
|||||
|
AAA1518009
|
Thermo Scientific Chemicals
A1518009 |
10 g |
Each for $121.81
|
|
|||||
|
AAA1518022
|
Thermo Scientific Chemicals
A1518022 |
100 g |
Each for $720.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsWidely used as preservative, water treatment in agriculture. Also used as antistatic agent in textiles, fibers, leather, as flocculants, deodorant and as phase transfer catalyst.
Solubility
Soluble in ethanol. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
| 14866-43-4 | |
| 567.64 | |
| UXMZNEHSMYESLH-UHFFFAOYSA-M | |
| 3084210 | |
| [Br-].CCCCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C34H48BrP | |
| MFCD00051858 | |
| hexadecyltriphenylphosphonium bromide, 1-hexadecyl triphenylphosphonium bromide, cetyltriphenylphosphonium bromide, phosphonium, hexadecyltriphenyl-, bromide, hexadecyl triphenyl phosphanium bromide, hexadecyltriphenylphosphanium bromide, acmc-1boiz, hexadecyl triphenyl phosphonium bromide, hexadecyl triphenyl phosphonium chloride, 1-hexadecyl triphenylphosphoniumbromide | |
| hexadecyl(triphenyl)phosphanium;bromide |
Specifications
| 14866-43-4 | |
| C34H48BrP | |
| 5 g | |
| Hygroscopic | |
| Soluble in ethanol. Slightly soluble in water. | |
| [Br-].CCCCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 567.64 | |
| 567.64 | |
| (1-Hexadecyl)triphenylphosphonium bromide |
| 99°C to 101°C | |
| MFCD00051858 | |
| 3582592 | |
| hexadecyltriphenylphosphonium bromide, 1-hexadecyl triphenylphosphonium bromide, cetyltriphenylphosphonium bromide, phosphonium, hexadecyltriphenyl-, bromide, hexadecyl triphenyl phosphanium bromide, hexadecyltriphenylphosphanium bromide, acmc-1boiz, hexadecyl triphenyl phosphonium bromide, hexadecyl triphenyl phosphonium chloride, 1-hexadecyl triphenylphosphoniumbromide | |
| UXMZNEHSMYESLH-UHFFFAOYSA-M | |
| hexadecyl(triphenyl)phosphanium;bromide | |
| 3084210 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-939-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only