missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexadecyl)triphenylphosphonium bromide, 98+%

Catalog No. AAA1518006
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1518006 5 g
AAA1518009 10 g
AAA1518022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1518006 Supplier Thermo Scientific Chemicals Supplier No. A1518006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14866-43-4 | C34H48BrP | 567.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Widely used as preservative, water treatment in agriculture. Also used as antistatic agent in textiles, fibers, leather, as flocculants, deodorant and as phase transfer catalyst.

Solubility
Soluble in ethanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 14866-43-4
Molecular Formula C34H48BrP
Molecular Weight (g/mol) 567.64
MDL Number MFCD00051858
InChI Key UXMZNEHSMYESLH-UHFFFAOYSA-M
Synonym hexadecyltriphenylphosphonium bromide, 1-hexadecyl triphenylphosphonium bromide, cetyltriphenylphosphonium bromide, phosphonium, hexadecyltriphenyl-, bromide, hexadecyl triphenyl phosphanium bromide, hexadecyltriphenylphosphanium bromide, acmc-1boiz, hexadecyl triphenyl phosphonium bromide, hexadecyl triphenyl phosphonium chloride, 1-hexadecyl triphenylphosphoniumbromide
PubChem CID 3084210
IUPAC Name hexadecyl(triphenyl)phosphanium;bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 99°C to 101°C
Quantity 5 g
Beilstein 3582592
Sensitivity Hygroscopic
Solubility Information Soluble in ethanol. Slightly soluble in water.
Formula Weight 567.64
Percent Purity ≥98%
Chemical Name or Material (1-Hexadecyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-939-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only