Learn More
(1-Hexadecyl)trimethylammonium bromide, 98%
CAS: 57-09-0 | C19H42BrN | 364.46 g/mol
Supplier: Thermo Scientific Chemicals A152350I
Description
(1-Hexadecyl)trimethylammonium bromide is a cationic surfactant in hair conditioning products. It acts as a phase-transfer catalyst for the preparation of polyamide, polycarbonate and polythiocarbonate. It facilitates ligand-free palladium salt-catalyzed Heck and Suzuki cross-coupling reactions in water. It is an active component of the topical antiseptic cetrimide and used in household products such as shampoos and cosmetics. As a buffer solution, it is involved in the extraction of DNA as well as in the synthesis of gold nanoparticles and silican nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1-Hexadecyl)trimethylammonium bromide is a cationic surfactant in hair conditioning products. It acts as a phase-transfer catalyst for the preparation of polyamide, polycarbonate and polythiocarbonate. It facilitates ligand-free palladium salt-catalyzed Heck and Suzuki cross-coupling reactions in water. It is an active component of the topical antiseptic cetrimide and used in household products such as shampoos and cosmetics. As a buffer solution, it is involved in the extraction of DNA as well as in the synthesis of gold nanoparticles and silican nanoparticles.
Solubility
Soluble in alcohol, chloroform and water. Slightly soluble in acetone. Insoluble in diethyl ether and benzene.
Notes
Hygroscopic. Store in an airtight container. Incompatible with bases and strong oxidizing agents.
Specifications
| (1-Hexadecyl)trimethylammonium bromide | |
| ∼245°C (decomposition) | |
| 244°C (471°F) | |
| C19H42BrN | |
| 5000 g | |
| 3598189 | |
| 14,2025 | |
| Soluble in alcohol,chloroform and water. Slightly soluble in acetone. Insoluble in diethyl ether and benzene. | |
| [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C | |
| 364.46 | |
| CHEBI:3567 | |
| 98% |
| 57-09-0 | |
| 5 to 7 | |
| Odorless | |
| MFCD00011772 | |
| UN3077 | |
| Hygroscopic | |
| cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon | |
| LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
| hexadecyltrimethylazanium bromide | |
| 5974 | |
| 364.46 |
Chemical Identifiers
| 57-09-0 | |
| 364.46 | |
| LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
| 5974 | |
| [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
| C19H42BrN | |
| MFCD00011772 | |
| cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon | |
| CHEBI:3567 |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P362-P501c
H302-H315-H318-H335-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 200-311-3
RTECSNumber : BQ7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only