missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecene 99.0+%, TCI America™

Catalog No. p-7130632
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
H061025ML 25 mL
H0610100ML 100 mL
H0610500ML 500 mL
3 options

Catalog No. H061025ML

Supplier: TCI America H061025ML

Only null left

Chemical Identifiers

CAS 629-73-2
Molecular Formula C16H32
Molecular Weight (g/mol) 224.432
MDL Number MFCD00008991
InChI Key GQEZCXVZFLOKMC-UHFFFAOYSA-N
Synonym 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
PubChem CID 12395
ChEBI CHEBI:77507
IUPAC Name hexadec-1-ene
SMILES CCCCCCCCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 285°C
Quantity 25 mL
Formula Weight 224.43
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexadecene
EINECSNumber (2)-0027
RTECSNumber MM0479000
TSCA Yes