missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecene 99.0+%, TCI America™

Catalog No. H0610100ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0610100ML 100 mL
H061025ML 25 mL
H0610500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0610100ML Supplier TCI America Supplier No. H0610100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-73-2
Molecular Formula C16H32
Molecular Weight (g/mol) 224.432
MDL Number MFCD00008991
InChI Key GQEZCXVZFLOKMC-UHFFFAOYSA-N
Synonym 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
PubChem CID 12395
ChEBI CHEBI:77507
IUPAC Name hexadec-1-ene
SMILES CCCCCCCCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 285°C
Quantity 100 mL
Formula Weight 224.43
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexadecene
EINECSNumber (2)-0027
RTECSNumber MM0479000
TSCA Yes