missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecene 90.0+%, TCI America™

Catalog No. p-7130485
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
H032325ML 25 mL
H0323500ML 500 mL
2 options

Catalog No. H032325ML

Supplier: TCI America H032325ML

Only null left

Chemical Identifiers

CAS 629-73-2
Molecular Formula C16H32
Molecular Weight (g/mol) 224.432
MDL Number MFCD00008991
InChI Key GQEZCXVZFLOKMC-UHFFFAOYSA-N
Synonym 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
PubChem CID 12395
ChEBI CHEBI:77507
IUPAC Name hexadec-1-ene
SMILES CCCCCCCCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 285°C
Quantity 25 mL
Formula Weight 224.43
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexadecene
EINECSNumber (2)-0027
RTECSNumber MM0479000
TSCA Yes