Learn More
1-Hexadecene, 90+%
CAS: 629-73-2 | C16H32 | 224.432 g/mol
$266.56 - $266.56
Chemical Identifiers
| CAS | 629-73-2 |
|---|---|
| Molecular Formula | C16H32 |
| Molecular Weight (g/mol) | 224.432 |
| MDL Number | MFCD00008991 |
| InChI Key | GQEZCXVZFLOKMC-UHFFFAOYSA-N |
| Synonym | 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene |
| PubChem CID | 12395 |
| ChEBI | CHEBI:77507 |
| IUPAC Name | hexadec-1-ene |
| SMILES | CCCCCCCCCCCCCCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL02637AU
|
Thermo Scientific Chemicals
L02637AU |
1000 mL |
Each for $266.56
|
|
|||||
Description
1-Hexadecene is used as an intermediate for organic synthesis. It is utilized as a blend for off-shore drilling fluids. It is also used as a surface active agent, finishing agent, process regulator, functional fluid, lubricant and lubricant additive. Further, it acts as comonomers in polymer such as low density polyethylenes, which shows thiner, flexibility and tearing resistance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Hexadecene is used as an intermediate for organic synthesis. It is utilized as a blend for off-shore drilling fluids. It is also used as a surface active agent, finishing agent, process regulator, functional fluid, lubricant and lubricant additive. Further, it acts as comonomers in polymer such as low density polyethylenes, which shows thiner, flexibility and tearing resistance.
Solubility
Miscible with ether, petroleum ether and alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 629-73-2 | |
| 224.432 | |
| GQEZCXVZFLOKMC-UHFFFAOYSA-N | |
| 12395 | |
| hexadec-1-ene |
| C16H32 | |
| MFCD00008991 | |
| 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene | |
| CHEBI:77507 | |
| CCCCCCCCCCCCCCC=C |
Specifications
| 629-73-2 | |
| 0.781 | |
| 132°C (269°F) | |
| 1.442 | |
| 1000 mL | |
| 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene | |
| GQEZCXVZFLOKMC-UHFFFAOYSA-N | |
| hexadec-1-ene | |
| 12395 | |
| 224.43 | |
| 1-Hexadecene |
| 4°C | |
| 273°C to 274°C | |
| C16H32 | |
| MFCD00008991 | |
| 1753401 | |
| Miscible with ether,petroleum ether and alcohol. Immiscible with water. | |
| CCCCCCCCCCCCCCC=C | |
| 224.432 | |
| CHEBI:77507 | |
| 94% |
Safety and Handling
P301+P310-P331-P501c
H304
EINECSNumber : 211-105-8
RTECSNumber : MM0479000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only