missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanol 98.0+%, TCI America™

Catalog No. p-7130318
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H007125G 25 g
H0071500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H007125G Supplier TCI America Supplier No. H007125G
Only null left

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
PubChem CID 2682
ChEBI CHEBI:16125
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

Melting Point 50°C
Color White
Boiling Point 344°C
Quantity 25 g
Formula Weight 242.45
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hexadecanol
EINECSNumber (2)-0217
RTECSNumber MM0225000
TSCA Yes