missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanethiol 97.0+%, TCI America™

Catalog No. H0068500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0068500ML 500 mL
H006825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0068500ML Supplier TCI America Supplier No. H0068500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2917-26-2
Molecular Formula C16H34S
Molecular Weight (g/mol) 258.508
MDL Number MFCD00011677
InChI Key ORTRWBYBJVGVQC-UHFFFAOYSA-N
Synonym 1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptan
PubChem CID 18015
IUPAC Name hexadecane-1-thiol
SMILES CCCCCCCCCCCCCCCCS

Specifications

Color Yellow
Boiling Point 189°C
Quantity 500 mL
Formula Weight 258.51
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexadecanethiol
EINECSNumber (2)-0464
TSCA Yes