accessibility menu, dialog, popup

UserName

H3CSH

CAS RN 2917-26-2

IUPAC Name: hexadecane-1-thiol
Synonyms: 1-hexadecanethiol
Molecular Weight (g/mol): 258.51
Molecular Formula: C16H34S
InChi Key: ORTRWBYBJVGVQC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCS
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)