missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanethiol, 97% (dry wt.), may cont. up to 4% water

Catalog No. AAL15099AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15099AC 25 mL
AAL15099AE 100 mL
AAL15099AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL15099AC Supplier Thermo Scientific Chemicals Supplier No. L15099AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2917-26-2 | C16H34S | 258.508 g/mol

1-Hexadecanethiol is used in the study of nanoparticles and self-assembled monolayers. It is also used to prepare thiol-functionalized 1.5 nm gold nanoparticles via a ligand exchange process with triphenylphosphine-stabilized nanoparticles and in the formation of hydrophobic self-assembled monolayers (SAMs). Due to the length of the alkane chain, the resulting monolayer is highly ordered. 1-Hexadecanethiol is also employed in different fields such as biomedicine, chemical industry, also used in electroplating industry and chemical auxiliaries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecanethiol is used in the study of nanoparticles and self-assembled monolayers. It is also used to prepare thiol-functionalized 1.5 nm gold nanoparticles via a ligand exchange process with triphenylphosphine-stabilized nanoparticles and in the formation of hydrophobic self-assembled monolayers (SAMs). Due to the length of the alkane chain, the resulting monolayer is highly ordered. 1-Hexadecanethiol is also employed in different fields such as biomedicine, chemical industry, also used in electroplating industry and chemical auxiliaries.

Solubility
Soluble in methanol, benzene, ether, acetone and ethyl acetate. Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2917-26-2
Molecular Formula C16H34S
Molecular Weight (g/mol) 258.508
MDL Number MFCD00011677
InChI Key ORTRWBYBJVGVQC-UHFFFAOYSA-N
Synonym 1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptan
PubChem CID 18015
IUPAC Name hexadecane-1-thiol
SMILES CCCCCCCCCCCCCCCCS

Specifications

Melting Point 19°C to 23°C
Density 0.846
Boiling Point 334°C
Flash Point 135°C (275°F)
Odor Unpleasant
Refractive Index 1.4625
Linear Formula CH3(CH2)15SH
Quantity 25 mL
Beilstein 1748495
Sensitivity Air Sensitive
Solubility Information Soluble in methanol,benzene,ether,acetone and ethyl acetate. Not miscible or difficult to mix in water.
Formula Weight 258.51
Percent Purity 97%
Assay (dry wt.), may cont. up to 4% water
Chemical Name or Material 1-Hexadecanethiol
Show More Show Less
EINECSNumber 220-846-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only