missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexacosanol 95.0+%, TCI America™
$290.79 - $1224.68
Chemical Identifiers
| CAS | 506-52-5 |
|---|---|
| Molecular Formula | C26H54O |
| Molecular Weight (g/mol) | 382.717 |
| MDL Number | MFCD00002940 |
| InChI Key | IRHTZOCLLONTOC-UHFFFAOYSA-N |
| Synonym | 1-hexacosanol, ceryl alcohol, n-hexacosanol, hexacosyl alcohol, hexacosanol, cerylic alcohol, unii-m7sd300nnb, m7sd300nnb, cerylalkohol, hexacosanyl alcohol |
| PubChem CID | 68171 |
| ChEBI | CHEBI:28415 |
| IUPAC Name | hexacosan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H0342100MG
|
TCI America
H0342100MG |
100 mg |
Each for $290.79
|
|
|||||
|
H03421G
|
TCI America
H03421G |
1 g |
Each for $1,224.68
|
|
|||||
Chemical Identifiers
| 506-52-5 | |
| 382.717 | |
| IRHTZOCLLONTOC-UHFFFAOYSA-N | |
| 68171 | |
| hexacosan-1-ol |
| C26H54O | |
| MFCD00002940 | |
| 1-hexacosanol, ceryl alcohol, n-hexacosanol, hexacosyl alcohol, hexacosanol, cerylic alcohol, unii-m7sd300nnb, m7sd300nnb, cerylalkohol, hexacosanyl alcohol | |
| CHEBI:28415 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCO |
Specifications
| 506-52-5 | |
| White-Yellow | |
| MFCD00002940 | |
| 1-hexacosanol, ceryl alcohol, n-hexacosanol, hexacosyl alcohol, hexacosanol, cerylic alcohol, unii-m7sd300nnb, m7sd300nnb, cerylalkohol, hexacosanyl alcohol | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCO | |
| 382.717 | |
| CHEBI:28415 | |
| ≥95.0% (GC) | |
| 1-Hexacosanol |
| 80°C | |
| C26H54O | |
| 100 mg | |
| IRHTZOCLLONTOC-UHFFFAOYSA-N | |
| hexacosan-1-ol | |
| 68171 | |
| 382.72 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes