accessibility menu, dialog, popup

UserName

CAS RN 506-52-5

H3COH

CAS RN 506-52-5

IUPAC Name: hexacosan-1-ol
Synonyms: ceryl alcohol cerylic alcohol
Molecular Weight (g/mol): 382.72
Molecular Formula: C26H54O
InChi Key: IRHTZOCLLONTOC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results