missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptyn-3-ol 97.0+%, TCI America™
$54.06 - $141.67
Chemical Identifiers
| CAS | 7383-19-9 |
|---|---|
| Molecular Formula | C7H12O |
| Molecular Weight (g/mol) | 112.172 |
| MDL Number | MFCD00041597 |
| InChI Key | SHSFXAVQBIEYMK-UHFFFAOYSA-N |
| Synonym | 1-heptyn-3-ol, n-butyl ethynyl carbinol, acmc-1biv8, 1-heptyn-3-ol, +/- |
| PubChem CID | 93050 |
| IUPAC Name | hept-1-yn-3-ol |
| SMILES | CCCCC(C#C)O |
Chemical Identifiers
| 7383-19-9 | |
| 112.172 | |
| SHSFXAVQBIEYMK-UHFFFAOYSA-N | |
| 93050 | |
| CCCCC(C#C)O |
| C7H12O | |
| MFCD00041597 | |
| 1-heptyn-3-ol, n-butyl ethynyl carbinol, acmc-1biv8, 1-heptyn-3-ol, +/- | |
| hept-1-yn-3-ol |
Specifications
| 7383-19-9 | |
| 73°C | |
| MFCD00041597 | |
| 1-heptyn-3-ol, n-butyl ethynyl carbinol, acmc-1biv8, 1-heptyn-3-ol, +/- | |
| CCCCC(C#C)O | |
| 112.172 | |
| 112.17 | |
| Liquid |
| Yellow | |
| C7H12O | |
| 1 mL | |
| SHSFXAVQBIEYMK-UHFFFAOYSA-N | |
| hept-1-yn-3-ol | |
| 93050 | |
| ≥97.0% (GC) | |
| 1-Heptyn-3-ol |
Safety and Handling
TSCA : No