accessibility menu, dialog, popup

UserName

H3COHCH

CAS RN 7383-19-9

IUPAC Name: hept-1-yn-3-ol
Synonyms: hept-1-yn-3-ol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: SHSFXAVQBIEYMK-UHFFFAOYNA-N
SMILES: CCCCC(O)C#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results