missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptyn-3-ol 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 73°C |
| Quantity | 5 mL |
| Formula Weight | 112.17 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Heptyn-3-ol |
Safety and Handling
| TSCA | No |