missing translation for 'onlineSavingsMsg'
Learn More

1-Heptylamine, 98+%

Catalog No. aaa1792218
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
50 g
Catalog No. Quantity
AAA1792218 50 g
AAA1792214 25 g
AAA1792230 250 g
3 options

Catalog No. AAA1792218

Supplier: Thermo Scientific Chemicals A1792218

Only null left

CAS: 111-68-2 | C7H17N | 115.22 g/mol

1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

Solubility
Soluble in water 6791 mg/L @ 25°C.

Notes
Air sensitive. Store away from oxidizing agents, air, acids.

Chemical Identifiers

CAS 111-68-2
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008244
InChI Key WJYIASZWHGOTOU-UHFFFAOYSA-N
Synonym heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681
PubChem CID 8127
IUPAC Name heptan-1-amine
SMILES CCCCCCCN

Specifications

Melting Point -23°C
Density 0.775
Boiling Point 154°C to 156°C
Flash Point 35°C (95°F)
Odor Amine-like
Refractive Index 1.4245
Linear Formula CH3(CH2)6NH2
Quantity 50 g
UN Number UN2734
Beilstein 1731688
Sensitivity Air Sensitive
Solubility Information Soluble in water 6791mg/L @ 25°C.
Formula Weight 115.22
Percent Purity ≥98%
Chemical Name or Material 1-Heptylamine
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 203-895-8
RTECSNumber MK0600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only