missing translation for 'onlineSavingsMsg'
Learn More

1-Heptene 98.0+%, TCI America™

Catalog No. p-7130306
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H004225ML 25 mL
H0042100ML 100 mL
H0042500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H004225ML Supplier TCI America Supplier No. H004225ML
Only null left

Chemical Identifiers

CAS 592-76-7
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00009531
InChI Key ZGEGCLOFRBLKSE-UHFFFAOYSA-N
Synonym 1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum
PubChem CID 11610
IUPAC Name hept-1-ene
SMILES CCCCCC=C

Specifications

Color Colorless
Boiling Point 94°C
Quantity 25 mL
UN Number 2278
Formula Weight 98.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Heptene
EINECSNumber (2)-2359
RTECSNumber MJ8840000
TSCA Yes