accessibility menu, dialog, popup

UserName

CAS RN 592-76-7

IUPAC Name: hept-1-ene
Synonyms: heptene
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: ZGEGCLOFRBLKSE-UHFFFAOYSA-N
SMILES: CCCCCC=C
Boiling Point 
  • (2)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (6)