missing translation for 'onlineSavingsMsg'
Learn More

1-Heptene, 98+%

Catalog No. AAL11487AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11487AC 25 mL
AAL11487AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL11487AC Supplier Thermo Scientific Chemicals Supplier No. L11487AC

CAS: 592-76-7 | C7H14 | 98.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 592-76-7
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00009531
InChI Key ZGEGCLOFRBLKSE-UHFFFAOYSA-N
Synonym 1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum
PubChem CID 11610
IUPAC Name hept-1-ene
SMILES CCCCCC=C

Specifications

Melting Point ∼-119°C
Density 0.697
Boiling Point 93°C to 94°C
Flash Point −8°C (17°F)
Refractive Index 1.4
Linear Formula CH3(CH2)4CH=CH2
Quantity 25 mL
UN Number UN2278
Beilstein 1098332
Formula Weight 98.19
Percent Purity ≥98%
Chemical Name or Material 1-Heptene
Show More Show Less
Hazard Category H225-H304-H315-H319
Hazard Statement GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-HEPTENE
EINECSNumber 209-767-8
RTECSNumber MJ8840000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only