Learn More
1-Heptene, 98+%, Thermo Scientific Chemicals
$158.70 - $427.73
Chemical Identifiers
CAS | 592-76-7 |
---|---|
Molecular Formula | C7H14 |
Molecular Weight (g/mol) | 98.189 |
MDL Number | MFCD00009531 |
InChI Key | ZGEGCLOFRBLKSE-UHFFFAOYSA-N |
Synonym | 1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum |
PubChem CID | 11610 |
IUPAC Name | hept-1-ene |
SMILES | CCCCCC=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL11487AC
|
Thermo Scientific Chemicals
L11487AC |
25 mL |
Each for $158.70
|
|
|||||
AAL11487AE
|
Thermo Scientific Chemicals
L11487AE |
100 mL |
Each for $427.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
592-76-7 | |
98.189 | |
ZGEGCLOFRBLKSE-UHFFFAOYSA-N | |
11610 | |
CCCCCC=C |
C7H14 | |
MFCD00009531 | |
1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum | |
hept-1-ene |
Specifications
592-76-7 | |
0.697 | |
−8°C (17°F) | |
1.4 | |
MFCD00009531 | |
UN2278 | |
1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum | |
CCCCCC=C | |
98.189 | |
98.19 | |
1-Heptene |
∼-119°C | |
93°C to 94°C | |
C7H14 | |
CH3(CH2)4CH=CH2 | |
25 mL | |
1098332 | |
ZGEGCLOFRBLKSE-UHFFFAOYSA-N | |
hept-1-ene | |
11610 | |
≥98% |
Safety and Handling
GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-HEPTENE
EINECSNumber : 209-767-8
RTECSNumber : MJ8840000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only