missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Heptafluorobutyryl)imidazole 97.0+%, TCI America™
$205.05 - $833.24
Chemical Identifiers
| CAS | 32477-35-3 |
|---|---|
| Molecular Formula | C7H3F7N2O |
| Molecular Weight (g/mol) | 264.103 |
| MDL Number | MFCD00014503 |
| InChI Key | MSYHGYDAVLDKCE-UHFFFAOYSA-N |
| Synonym | n-heptafluorobutyrylimidazole, 1-heptafluorobutyryl imidazole, heptafluorobutyrylimidazole, n-heptafluorobutyryl imidazole, hfbi, 1-perfluorobutyryl imidazole, 2,2,3,3,4,4,4-heptafluoro-1-1h-imidazol-1-yl butan-1-one, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl-1h-imidazole, 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-yl butan-1-one, 1h-imidazole, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl |
| PubChem CID | 94431 |
| IUPAC Name | 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one |
| SMILES | C1=CN(C=N1)C(=O)C(C(C(F)(F)F)(F)F)(F)F |
Chemical Identifiers
| 32477-35-3 | |
| 264.103 | |
| MSYHGYDAVLDKCE-UHFFFAOYSA-N | |
| 94431 | |
| C1=CN(C=N1)C(=O)C(C(C(F)(F)F)(F)F)(F)F |
| C7H3F7N2O | |
| MFCD00014503 | |
| n-heptafluorobutyrylimidazole, 1-heptafluorobutyryl imidazole, heptafluorobutyrylimidazole, n-heptafluorobutyryl imidazole, hfbi, 1-perfluorobutyryl imidazole, 2,2,3,3,4,4,4-heptafluoro-1-1h-imidazol-1-yl butan-1-one, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl-1h-imidazole, 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-yl butan-1-one, 1h-imidazole, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl | |
| 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one |
Specifications
| 32477-35-3 | |
| Yellow | |
| C7H3F7N2O | |
| 5 g | |
| MSYHGYDAVLDKCE-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one | |
| 94431 | |
| ≥97.0% (GC,T) | |
| 1-(Heptafluorobutyryl)imidazole [Acylating Agent] |
| 13°C | |
| 161°C | |
| MFCD00014503 | |
| n-heptafluorobutyrylimidazole, 1-heptafluorobutyryl imidazole, heptafluorobutyrylimidazole, n-heptafluorobutyryl imidazole, hfbi, 1-perfluorobutyryl imidazole, 2,2,3,3,4,4,4-heptafluoro-1-1h-imidazol-1-yl butan-1-one, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl-1h-imidazole, 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-yl butan-1-one, 1h-imidazole, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl | |
| C1=CN(C=N1)C(=O)C(C(C(F)(F)F)(F)F)(F)F | |
| 264.103 | |
| 264.10 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator