missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptadecanol, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 1454-85-9 |
|---|---|
| Molecular Formula | C17H36O |
| Molecular Weight (g/mol) | 256.474 |
| InChI Key | GOQYKNQRPGWPLP-UHFFFAOYSA-N |
| Synonym | 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1 |
| PubChem CID | 15076 |
| ChEBI | CHEBI:77470 |
| IUPAC Name | heptadecan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC221230050
|
Thermo Scientific Chemicals
221230050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC221230250
|
Thermo Scientific Chemicals
221230250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1454-85-9 | |
| 256.474 | |
| 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1 | |
| CHEBI:77470 | |
| CCCCCCCCCCCCCCCCCO |
| C17H36O | |
| GOQYKNQRPGWPLP-UHFFFAOYSA-N | |
| 15076 | |
| heptadecan-1-ol |
Specifications
| 1454-85-9 | |
| 154°C | |
| 96% min. (GC) | |
| C17H36O | |
| 5g | |
| 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1 | |
| GOQYKNQRPGWPLP-UHFFFAOYSA-N | |
| heptadecan-1-ol | |
| 15076 | |
| 256.46 | |
| 1-Heptadecanol |
| 308.5°C | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)16OH | |
| 01,I,22 | |
| Solubility in water: slightly soluble in water. | |
| CCCCCCCCCCCCCCCCCO | |
| 256.46 | |
| CHEBI:77470 | |
| 97% |
Safety and Handling
EINECSNumber : 215-932-5
RUO â Research Use Only