missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptadecanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 221230250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 1-Heptadecanol | |
| 308.5°C | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)16OH | |
| 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1 | |
| GOQYKNQRPGWPLP-UHFFFAOYSA-N | |
| heptadecan-1-ol | |
| 15076 | |
| 256.46 |
| 1454-85-9 | |
| 154°C | |
| 97% | |
| C17H36O | |
| 25g | |
| Solubility in water: slightly soluble in water. | |
| CCCCCCCCCCCCCCCCCO | |
| 256.474 | |
| CHEBI:77470 | |
| 97% |
Chemical Identifiers
| 1454-85-9 | |
| 256.474 | |
| 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1 | |
| CHEBI:77470 | |
| CCCCCCCCCCCCCCCCCO |
| C17H36O | |
| GOQYKNQRPGWPLP-UHFFFAOYSA-N | |
| 15076 | |
| heptadecan-1-ol |
RUO â Research Use Only