missing translation for 'onlineSavingsMsg'
Learn More

1-Fmoc-piperidine-4-carboxylic acid, 98%

Catalog No. p-7055354
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6391506 5 g
AAH6391514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6391506 Supplier Thermo Scientific Chemicals Supplier No. H6391506

CAS: 148928-15-8 | C21H21NO4 | 351.402 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148928-15-8
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD00273475
InChI Key OJYOOHSQOIDDOO-UHFFFAOYSA-N
Synonym fmoc-inp-oh, fmoc-isonipecotic acid, 1-fmoc-piperidine-4-carboxylic acid, 1-9h-fluoren-9-ylmethoxy carbonyl piperidine-4-carboxylic acid, n-fmoc-isonipecotic acid, fmoc-piperidine-4-carboxylic acid, n-fmoc-4-piperidinecarboxylic acid, 1,4-piperidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl ester, n-9-fluorenylmethyloxycarbonyl-isonipecotic acid, 1-9h-fluoren-9-ylmethoxycarbonyl piperidine-4-carboxylic acid
PubChem CID 2736490
IUPAC Name 1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-4-carboxylic acid
SMILES C1CN(CCC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point ∼190°C (decomposition)
Color White
Quantity 5 g
Beilstein 7722550
Formula Weight 351.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Fmoc-piperidine-4-carboxylic acid
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only