missing translation for 'onlineSavingsMsg'
Learn More

1-(Fmoc-amino)cyclohexanecarboxylic acid, 98+%

Catalog No. AAL1981006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1981006 5 g
AAL1981003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1981006 Supplier Thermo Scientific Chemicals Supplier No. L1981006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 162648-54-6 | C22H23NO4 | 365.43 g/mol

1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 162648-54-6
Molecular Formula C22H23NO4
Molecular Weight (g/mol) 365.43
MDL Number MFCD00273464
InChI Key VCIVAWBKUQJNSX-UHFFFAOYSA-N
Synonym 1-fmoc-amino cyclohexanecarboxylic acid, fmoc-ac6c-oh, fmoc-homocycloleucine, 1-9h-fluoren-9-yl methoxy carbonyl amino cyclohexanecarboxylic acid, fmoc-1-aminocyclohexanecarboxylic acid, 1-9h-fluoren-9-ylmethoxy carbonyl amino cyclohexane-1-carboxylic acid, acmc-209dor, fmoc-nh 1 chex-oh, 1-9h-fluoren-9-ylmethoxycarbonylamino cyclohexane-1-carboxylic acid
PubChem CID 2756093
IUPAC Name 1-(9H-fluoren-9-ylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
SMILES OC(=O)C1(CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 185°C to 187°C (decomposition)
Quantity 5 g
Beilstein 7887646
Solubility Information Insoluble in water.
Formula Weight 365.43
Percent Purity ≥98%
Chemical Name or Material 1-(Fmoc-amino)cyclohexanecarboxylic acid
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only