missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. p-7129760
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05695G 5 g
F056925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05695G Supplier TCI America Supplier No. F05695G

Chemical Identifiers

CAS 1077-01-6
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.102
MDL Number MFCD00236323
InChI Key AUKDFDQPJWJEDH-UHFFFAOYSA-N
PubChem CID 2777286
IUPAC Name 1-fluoro-3-(trifluoromethoxy)benzene
SMILES C1=CC(=CC(=C1)F)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 105°C
Quantity 5 g
UN Number 1993
Formula Weight 180.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Fluoro-3-(trifluoromethoxy)benzene