missing translation for 'onlineSavingsMsg'
Learn More

1-Fluorenecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129742
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05411G 1 g
F05415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05411G Supplier TCI America Supplier No. F05411G
Only null left

Chemical Identifiers

CAS 6276-03-5
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00001130
InChI Key HTPXFGUCAUTOEL-UHFFFAOYSA-N
Synonym 1-fluorenecarboxylic acid, fluorene-1-carboxylic acid, fluorenecarboxylic acid, unii-ao792vd660, 1-fluorene carboxylic acid, 9h-fluorene-1-carboxylicacid, htpxfgucautoel-uhfffaoysa
PubChem CID 80468
IUPAC Name 9H-fluorene-1-carboxylic acid
SMILES C1C2=CC=CC=C2C3=C1C(=CC=C3)C(=O)O

Specifications

Melting Point 248°C
Color Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 210.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Fluorenecarboxylic Acid