missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynylpyrene, 98%

Catalog No. p-7052581
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH37327MD 250 mg
AAH3732703 1 g
AAH3732706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH37327MD Supplier Thermo Scientific Chemicals Supplier No. H37327MD

CAS: 34993-56-1 | C18H10 | 226.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34993-56-1
Molecular Formula C18H10
Molecular Weight (g/mol) 226.28
MDL Number MFCD02093933
InChI Key VEBUBSLYGRMOSZ-UHFFFAOYSA-N
Synonym 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2
PubChem CID 154905
IUPAC Name 1-ethynylpyrene
SMILES C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 113°C to 116°C
Color Yellow
Quantity 250 mg
Beilstein 2365008
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 1-Ethynylpyrene

RUO – Research Use Only