missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethynylpyrene, 96%
CAS: 34993-56-1 | C18H10 | 226.28 g/mol
$660.22 - $660.22
Chemical Identifiers
| CAS | 34993-56-1 |
|---|---|
| Molecular Formula | C18H10 |
| Molecular Weight (g/mol) | 226.28 |
| MDL Number | MFCD02093933 |
| InChI Key | VEBUBSLYGRMOSZ-UHFFFAOYSA-N |
| Synonym | 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2 |
| PubChem CID | 154905 |
| IUPAC Name | 1-ethynylpyrene |
| SMILES | C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1722706
|
Thermo Scientific Chemicals
L1722706 |
5 g |
Each for $660.22
|
|
|||||
Description
1-Ethynylpyrene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Ethynylpyrene is used as a pharmaceutical intermediate.
Solubility
Soluble in chloroform, dichloromethane and toluene. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
| 34993-56-1 | |
| 226.28 | |
| VEBUBSLYGRMOSZ-UHFFFAOYSA-N | |
| 154905 | |
| C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 |
| C18H10 | |
| MFCD02093933 | |
| 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2 | |
| 1-ethynylpyrene |
Specifications
| 34993-56-1 | |
| C18H10 | |
| 5 g | |
| 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2 | |
| VEBUBSLYGRMOSZ-UHFFFAOYSA-N | |
| 1-ethynylpyrene | |
| 154905 | |
| 96% |
| 113°C to 116°C | |
| MFCD02093933 | |
| 2365008 | |
| Soluble in chloroform,dichloromethane and toluene. Insoluble in water. | |
| C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 | |
| 226.28 | |
| 226.28 | |
| 1-Ethynylpyrene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only