missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynylpyrene, 96%

Catalog No. AAL1722706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1722706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1722706 Supplier Thermo Scientific Chemicals Supplier No. L1722706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34993-56-1 | C18H10 | 226.28 g/mol

1-Ethynylpyrene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylpyrene is used as a pharmaceutical intermediate.

Solubility
Soluble in chloroform, dichloromethane and toluene. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 34993-56-1
Molecular Formula C18H10
Molecular Weight (g/mol) 226.28
MDL Number MFCD02093933
InChI Key VEBUBSLYGRMOSZ-UHFFFAOYSA-N
Synonym 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2
PubChem CID 154905
IUPAC Name 1-ethynylpyrene
SMILES C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 113°C to 116°C
Quantity 5 g
Beilstein 2365008
Solubility Information Soluble in chloroform,dichloromethane and toluene. Insoluble in water.
Formula Weight 226.28
Percent Purity 96%
Chemical Name or Material 1-Ethynylpyrene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only