missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-4-(trifluoromethyl)benzene 98.0+%, TCI America™

Catalog No. E06265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06265G 5 g
E06261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06265G Supplier TCI America Supplier No. E06265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 705-31-7
Molecular Formula C9H5F3
Molecular Weight (g/mol) 170.13
MDL Number MFCD01861903
InChI Key XTKBMZQCDBHHKY-UHFFFAOYSA-N
Synonym 1-ethynyl-4-trifluoromethyl benzene, 4'-trifluoromethylphenyl acetylene, 4-trifluoromethyl phenylacetylene, 4-ethynyl-alpha,alpha,alpha-trifluorotoluene, 4-ethynylbenzotrifluoride, 4-ethynyl-a,a,a-trifluorotoluene, 4-ethynyl trifluorotoluene, benzene, 1-ethynyl-4-trifluoromethyl, 1-ethynyl-4-trifluoromethyl-benzene, 4-ethynyl-1-trifluoromethyl benzene
PubChem CID 4348295
IUPAC Name 1-ethynyl-4-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=C(C=C1)C#C

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
UN Number 1993
Formula Weight 170.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethynyl-4-(trifluoromethyl)benzene
TSCA No
Recommended Storage Refrigerator