missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-4-fluorobenzene 98.0+%, TCI America™

Catalog No. F04701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04701G 1 g
F04705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04701G Supplier TCI America Supplier No. F04701G

Chemical Identifiers

CAS 766-98-3
Molecular Formula C8H5F
Molecular Weight (g/mol) 120.126
MDL Number MFCD00168823
InChI Key QXSWHQGIEKUBAS-UHFFFAOYSA-N
Synonym 4-fluorophenylacetylene, benzene, 1-ethynyl-4-fluoro, 1-ethynyl-4-fluoro-benzene, 4'-fluorophenyl acetylene, p-fluorophenyl acetylene, 4'-fluorophenylacetylene, pubchem16719, 4-fluorophenyl acetylene, 4-fluoro phenylacetylene
PubChem CID 522627
IUPAC Name 1-ethynyl-4-fluorobenzene
SMILES C#CC1=CC=C(C=C1)F

Specifications

Color White-Yellow
Boiling Point 56°C
Quantity 1 g
UN Number 1325
Formula Weight 120.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Ethynyl-4-fluorobenzene
TSCA No
Recommended Storage Refrigerator